SY3H3D682296 Synthonix Corporation

Sodium 4-nitrobenzoate

  • Cas Number : 3847-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46455.html

Safety Information

MSDS https://synthonix.com/46455.html
Availability details for
Quantity
  • Availability
  • Add to Cart