SY3H3D682297 Synthonix Corporation

Sodium 5-amino-2-hydroxybenzoate

  • MDL number No MDL found
  • Cas Number : 35589-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/46456.html

Safety Information

MSDS https://synthonix.com/46456.html
Availability details for
Quantity
  • Availability
  • Add to Cart