SY3H3D68229A Synthonix Corporation

Sodium 2-oxo-2-phenylethanesulfonate

  • Cas Number : 63738-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48706.html

Safety Information

MSDS https://synthonix.com/48706.html
Availability details for
Quantity
  • Availability
  • Add to Cart