SY3H3D68229B Synthonix Corporation

2-(4-Sulfamoylphenyl)acetic acid

  • MDL number MFCD07801100
  • Cas Number : 22958-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49361.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49361.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart