SY3H3D68229C Synthonix Corporation

Sodium 2-((2-amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethanolate dihydrate

  • MDL number MFCD28902495
  • Cas Number : 261506-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49479.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49479.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart