SY3H3D68229E Synthonix Corporation

Sodium 2-(5-(trifluoromethyl)pyridin-2-yl)acetate

  • MDL number No MDL found
  • Cas Number : 1956366-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49481.html

Safety Information

MSDS https://synthonix.com/49481.html
Availability details for
Quantity
  • Availability
  • Add to Cart