SY3H3D6822A5 Synthonix Corporation

Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50215.html

Safety Information

MSDS https://synthonix.com/50215.html
Availability details for
Quantity
  • Availability
  • Add to Cart