SY3H3D6822A6 Synthonix Corporation

Spiro[3.5]nonane-1,3-dione

  • MDL number No MDL found
  • Cas Number : 455264-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50286.html

Safety Information

MSDS https://synthonix.com/50286.html
Availability details for
Quantity
  • Availability
  • Add to Cart