SY3H3D6822A8 Synthonix Corporation

Propan-2-amine (S)-2-(((benzyloxy)carbonyl)amino)-3,3-dimethylbutanoate

  • Cas Number : 1621085-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57863.html

Safety Information

MSDS https://synthonix.com/57863.html
Availability details for
Quantity
  • Availability
  • Add to Cart