SY3H3D6822AA Synthonix Corporation

Sodium 3-oxo-3-phenylpropanoate

  • MDL number No MDL found
  • Cas Number : 7063-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52053.html

Safety Information

MSDS https://synthonix.com/52053.html
Availability details for
Quantity
  • Availability
  • Add to Cart