SY3H3D6822AD Synthonix Corporation

Sodium 2-((4-formylphenyl)(methyl)amino)ethanesulfonate

  • Cas Number : 56405-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 70
Country of Origin USA
Product Link https://synthonix.com/52767.html

Safety Information

MSDS https://synthonix.com/52767.html
Availability details for
Quantity
  • Availability
  • Add to Cart