SY3H3D6822AF Synthonix Corporation

Spiro[4.5]decane-7,9-dione

  • MDL number No MDL found
  • Cas Number : 82683-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53087.html

Safety Information

MSDS https://synthonix.com/53087.html
Availability details for
Quantity
  • Availability
  • Add to Cart