SY3H3D6822B1 Synthonix Corporation

Sodium ((4-aminophenyl)sulfonyl)(6-chloropyrazin-2-yl)amide

  • MDL number No MDL found
  • Cas Number : 23307-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53696.html

Safety Information

MSDS https://synthonix.com/53696.html
Availability details for
Quantity
  • Availability
  • Add to Cart