SY3H3D6822B4 Synthonix Corporation

disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)oxolan-2-yl]methyl phosphate heptahydrate

  • Cas Number : 849725-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55435.html

Safety Information

MSDS https://synthonix.com/55435.html
Availability details for
Quantity
  • Availability
  • Add to Cart