SY3H3D6822B5 Synthonix Corporation

1H-Spiro[furo[3,4-c]pyridine-3,4'-piperidin]-1-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 475152-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55446.html

Safety Information

MSDS https://synthonix.com/55446.html
Availability details for
Quantity
  • Availability
  • Add to Cart