SY3H3D6822B6 Synthonix Corporation

7H-Spiro[furo[3,4-b]pyridine-5,4'-piperidin]-7-one hydrochloride

  • Cas Number : 475152-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55447.html

Safety Information

MSDS https://synthonix.com/55447.html
Availability details for
Quantity
  • Availability
  • Add to Cart