SY3H3D6822B7 Synthonix Corporation

Spiro[piperidine-4,4'-pyrido[2,3-d][1,3]oxazin]-2'(1'H)-one hydrochloride

  • Cas Number : 1038866-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55457.html

Safety Information

MSDS https://synthonix.com/55457.html
Availability details for
Quantity
  • Availability
  • Add to Cart