SY3H3D6822BA Synthonix Corporation

Sodium (6R,7R)-7-amino-3-(((6-hydroxy-2-methyl-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 131257-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55744.html

Safety Information

MSDS https://synthonix.com/55744.html
Availability details for
Quantity
  • Availability
  • Add to Cart