SY3H3D6822BB Synthonix Corporation

3H-Spiro[isobenzofuran-1,4'-piperidin]-3-one

  • MDL number No MDL found
  • Cas Number : 37663-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55822.html

Safety Information

MSDS https://synthonix.com/55822.html
Availability details for
Quantity
  • Availability
  • Add to Cart