SY3H3D6822BE Synthonix Corporation

2H-Spiro[benzofuran-3,4'-piperidine]

  • MDL number No MDL found
  • Cas Number : 171-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/55974.html

Safety Information

MSDS https://synthonix.com/55974.html
Availability details for
Quantity
  • Availability
  • Add to Cart