SY3H3D6822C0 Synthonix Corporation

4,4'-Sulfonylbis(bromobenzene)

  • MDL number No MDL found
  • Cas Number : 2050-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56472.html

Safety Information

MSDS https://synthonix.com/56472.html
Availability details for
Quantity
  • Availability
  • Add to Cart