SY3H3D6822C5 Synthonix Corporation

Spiro[isochroman-1,4'-piperidine]

  • MDL number No MDL found
  • Cas Number : 180160-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57018.html

Safety Information

MSDS https://synthonix.com/57018.html
Availability details for
Quantity
  • Availability
  • Add to Cart