SY3H3D6822C6 Synthonix Corporation

1'H-Spiro[piperidine-4,3'-quinolin]-2'(4'H)-one hydrochloride

  • Cas Number : 1175358-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57419.html

Safety Information

MSDS https://synthonix.com/57419.html
Availability details for
Quantity
  • Availability
  • Add to Cart