SY3H3D6822C8 Synthonix Corporation

Spiro[isochroman-1,4'-piperidine] hydrochloride

  • MDL number No MDL found
  • Cas Number : 173943-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57424.html

Safety Information

MSDS https://synthonix.com/57424.html
Availability details for
Quantity
  • Availability
  • Add to Cart