SY3H3D6822CC Synthonix Corporation

Sodium 9,10-dioxo-9,10-dihydroanthracene-2,7-disulfonate

  • Cas Number : 853-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 80
Country of Origin USA
Product Link https://synthonix.com/58022.html

Safety Information

MSDS https://synthonix.com/58022.html
Availability details for
Quantity
  • Availability
  • Add to Cart