SY3H3D6822D0 Synthonix Corporation

Sodium 4,4'-(propane-2,2-diyl)diphenolate

  • MDL number No MDL found
  • Cas Number : 2444-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58550.html

Safety Information

MSDS https://synthonix.com/58550.html
Availability details for
Quantity
  • Availability
  • Add to Cart