SY3H3D6822D4 Synthonix Corporation

Sodium 4-butyl-3,5-dioxo-1,2-diphenylpyrazolidin-4-ide

  • MDL number No MDL found
  • Cas Number : 129-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59463.html

Safety Information

MSDS https://synthonix.com/59463.html
Availability details for
Quantity
  • Availability
  • Add to Cart