SY3H3D6822D5 Synthonix Corporation

Sodium 2-(3,3-bis(4-(dimethylamino)phenyl)ureido)acetate

  • Cas Number : 115871-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59464.html

Safety Information

MSDS https://synthonix.com/59464.html
Availability details for
Quantity
  • Availability
  • Add to Cart