SY3H3D6822E6 Synthonix Corporation

Tetrahydrocyclopenta[c]pyrrole-1,3(2H,3aH)-dione

  • MDL number No MDL found
  • Cas Number : 5763-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/39419.html

Safety Information

MSDS https://synthonix.com/39419.html
Availability details for
Quantity
  • Availability
  • Add to Cart