SY3H3D682305 Synthonix Corporation

1,3,4,6-Tetrakis(methoxymethyl)tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione

  • MDL number No MDL found
  • Cas Number : 17464-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39683.html

Safety Information

MSDS https://synthonix.com/39683.html
Availability details for
Quantity
  • Availability
  • Add to Cart