SY3H3D682307 Synthonix Corporation

2-(Thiophen-2-yl)ethyl 4-methylbenzenesulfonate

  • MDL number No MDL found
  • Cas Number : 40412-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/39703.html

Safety Information

MSDS https://synthonix.com/39703.html
Availability details for
Quantity
  • Availability
  • Add to Cart