SY3H3D68230A Synthonix Corporation

Triallyl benzene-1,2,4-tricarboxylate

  • MDL number No MDL found
  • Cas Number : 2694-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39747.html

Safety Information

MSDS https://synthonix.com/39747.html
Availability details for
Quantity
  • Availability
  • Add to Cart