SY3H3D68230D Synthonix Corporation

1H-1,2,3-Triazol-1-amine

  • MDL number No MDL found
  • Cas Number : 584-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/39768.html

Safety Information

MSDS https://synthonix.com/39768.html
Availability details for
Quantity
  • Availability
  • Add to Cart