SY3H3D682310 Synthonix Corporation

1,2,4-Thiadiazol-5-amine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1588440-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/39799.html

Safety Information

MSDS https://synthonix.com/39799.html
Availability details for
Quantity
  • Availability
  • Add to Cart