SY3H3D682311 Synthonix Corporation

2H-1,2,4-triazole-3-carbonitrile

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39810.html

Safety Information

MSDS https://synthonix.com/39810.html
Availability details for
Quantity
  • Availability
  • Add to Cart