SY3H3D682312 Synthonix Corporation

3-Thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one

  • MDL number No MDL found
  • Cas Number : 626-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39825.html

Safety Information

MSDS https://synthonix.com/39825.html
Availability details for
Quantity
  • Availability
  • Add to Cart