SY3H3D682318 Synthonix Corporation

3,4,5-Tribromopyrazole

  • Cas Number : 17635-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39895.html

Safety Information

MSDS https://synthonix.com/39895.html
Availability details for
Quantity
  • Availability
  • Add to Cart