SY3H3D68231D Synthonix Corporation

(1H-1,2,4-triazol-5-yl)methanol hydrochloride

  • MDL number No MDL found
  • Cas Number : 1195596-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39912.html

Safety Information

MSDS https://synthonix.com/39912.html
Availability details for
Quantity
  • Availability
  • Add to Cart