SY3H3D682321 Synthonix Corporation

(S)-1,1,1-Trifluoropropan-2-amine hydrochloride

  • Cas Number : 125353-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39920.html

Safety Information

MSDS https://synthonix.com/39920.html
Availability details for
Quantity
  • Availability
  • Add to Cart