SY3H3D682328 Synthonix Corporation

4,5,6-Trichloropyrimidin-2-amine

  • Cas Number : 51501-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40006.html

Safety Information

MSDS https://synthonix.com/40006.html
Availability details for
Quantity
  • Availability
  • Add to Cart