SY3H3D682332 Synthonix Corporation

3,3,3-Trifluoro-2-methylpropanoic acid

  • MDL number MFCD19228247
  • Cas Number : 381-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to Gray to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40172.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40172.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart