SY3H3D682334 Synthonix Corporation

1,1,1-Trifluoro-4-iodobutane

  • MDL number No MDL found
  • Cas Number : 461-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/40178.html

Safety Information

MSDS https://synthonix.com/40178.html
Availability details for
Quantity
  • Availability
  • Add to Cart