SY3H3D682335 Synthonix Corporation

Thiazol-5-ylmethanamine

  • MDL number No MDL found
  • Cas Number : 161805-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40190.html

Safety Information

MSDS https://synthonix.com/40190.html
Availability details for
Quantity
  • Availability
  • Add to Cart