SY3H3D682340 Synthonix Corporation

2,4,5,6-Tetrachloropyrimidine

  • Cas Number : 1780-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/40293.html

Safety Information

MSDS https://synthonix.com/40293.html
Availability details for
Quantity
  • Availability
  • Add to Cart