SY3H3D682342 Synthonix Corporation

5-(Trifluoromethyl)thiazol-2-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 174886-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40317.html

Safety Information

MSDS https://synthonix.com/40317.html
Availability details for
Quantity
  • Availability
  • Add to Cart