SY3H3D68234C Synthonix Corporation

(2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal

  • MDL number No MDL found
  • Cas Number : 24259-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40398.html

Safety Information

MSDS https://synthonix.com/40398.html
Availability details for
Quantity
  • Availability
  • Add to Cart