SY3H3D682397 Synthonix Corporation

2,3,6-Trichloropyridine

  • MDL number MFCD00463516
  • Cas Number : 6515-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale yellow - Greyish yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40529.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40529.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart