SY3H3D68239A Synthonix Corporation

2,3,5-Trichloropyridin-4-ol

  • MDL number No MDL found
  • Cas Number : 1970-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40546.html

Safety Information

MSDS https://synthonix.com/40546.html
Availability details for
Quantity
  • Availability
  • Add to Cart