SY3H3D6823A2 Synthonix Corporation

2,2,2-Trifluoro-1-(1H-pyrazol-4-yl)ethanone

  • Cas Number : 161957-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40608.html

Safety Information

MSDS https://synthonix.com/40608.html
Availability details for
Quantity
  • Availability
  • Add to Cart