SY3H3D6823A4 Synthonix Corporation

1H-Thieno[2,3-d]imidazol-2(3H)-one

  • MDL number No MDL found
  • Cas Number : 71309-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40779.html

Safety Information

MSDS https://synthonix.com/40779.html
Availability details for
Quantity
  • Availability
  • Add to Cart